accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15761-39-4

CH3H3CCH3OONHOO

CAS RN 15761-39-4

IUPAC Name: 1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: ZQEBQGAAWMOMAI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products