man

Test

CAS RN 15804-19-0

OHNNHO

CAS RN 15804-19-0

IUPAC Name: 1,2,3,4-tetrahydroquinoxaline-2,3-dione
Synonyms: quinoxalinedione
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: ABJFBJGGLJVMAQ-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2NC1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results