accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 158364-83-1

CH3FFFF

CAS RN 158364-83-1

IUPAC Name: 2-fluoro-4-methyl-1-(trifluoromethyl)benzene
Synonyms: 2-fluoro-4-methyl-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 178.13
Molecular Formula: C8H6F4
InChi Key: GWWHXSLLCSBGRO-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(C=C1)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products