CAS RN 16682-12-5
CAS RN 16682-12-5
IUPAC Name:
hydrogen (2R)-2,5-diaminopentanoic acid chloride
Synonyms:
hydrogen L-(-)-ornithine chloride
Molecular Weight (g/mol):
168.62
Molecular Formula:
C5H13ClN2O2
InChi Key:
GGTYBZJRPHEQDG-PGMHMLKASA-N
SMILES:
[H+].[Cl-].NCCC[C@@H](N)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar D-Ornithine hydrochloride, 98+%
CAS: 16682-12-5 Molecular Formula: C5H13ClN2O2 Molecular Weight (g/mol): 168.621 MDL Number: MFCD00012917 InChI Key: GGTYBZJRPHEQDG-PGMHMLKASA-N Synonym: d-ornithine monohydrochloride,d-ornithine hydrochloride,d-ornithine hcl,h-d-orn-oh.hcl,r-ornithine hydrochloride,d-ornithine, monohydrochloride,l---ornithine hydrochloride,r-2,5-diaminopentanoic acid hydrochloride,2r-2,5-diaminopentanoic acid hydrochloride,ornithine hcl PubChem CID: 443122 ChEBI: CHEBI:4221 IUPAC Name: (2R)-2,5-diaminopentanoic acid;hydrochloride SMILES: C(CC(C(=O)O)N)CN.Cl