CAS RN 1672-88-4
CAS RN 1672-88-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Furagin, MedChemExpress
MedChemExpress Furagin, nitrofurantoin analog, is an anti-bacterial agent.
