CAS RN 170379-51-8
CAS RN 170379-51-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
3
av
3
Resultat
1
3-Methyl-8-(2’-deoxy-Beta-D-ribofuranosyl)isoxanthopterin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Methyl-8-(2’-deoxy-Beta-D-ribofuranosyl)isoxanthopterin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Methyl-8-(2’-deoxy-Beta-D-ribofuranosyl)isoxanthopterin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.