CAS RN 17102-63-5
CAS RN 17102-63-5
IUPAC Name:
(4-bromo-2-methoxyphenyl)methanol
Synonyms:
(4-bromo-2-methoxyphenyl)methanol
Molecular Weight (g/mol):
217.06
Molecular Formula:
C8H9BrO2
InChi Key:
HBEIHPSICGGZIF-UHFFFAOYSA-N
SMILES:
COC1=C(CO)C=CC(Br)=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4-Bromo-2-methoxybenzyl alcohol, 97%, Thermo Scientific™
CAS: 17102-63-5 Molecular Formula: C8H9BrO2 Molecular Weight (g/mol): 217.06 MDL Number: MFCD07369762 InChI Key: HBEIHPSICGGZIF-UHFFFAOYSA-N PubChem CID: 15128243 IUPAC Name: (4-bromo-2-methoxyphenyl)methanol SMILES: COC1=C(CO)C=CC(Br)=C1