accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 177842-14-7

H2NOFF

CAS RN 177842-14-7

IUPAC Name: 1-[4-(difluoromethoxy)phenyl]methanamine
Synonyms: 1-[4-(difluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 173.16
Molecular Formula: C8H9F2NO
InChi Key: UHWPLNHRROQPMB-UHFFFAOYSA-N
SMILES: NCC1=CC=C(OC(F)F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products