CAS RN 1796641-10-5
CAS RN 1796641-10-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Parimifasor, MedChemExpress
MedChemExpress Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity.
