CAS RN 1895895-38-1
CAS RN 1895895-38-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express CHZ868, MedChemExpress
MedChemExpress CHZ868 is a type II JAK2 inhibitor with an IC50 of 0.17 μM in EPOR JAK2 WT Ba/F3 cell.
