CAS RN 1919820-28-2
CAS RN 1919820-28-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Gcase activator 1, MedChemExpress
MedChemExpress Gcase activator 1 is an activator of glucocerebrosidase (Gcase) extracted from patent WO 2017192841 A1.
