accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 192575-19-2

CH3NNHNSOOOSOOOOOOPOOONaNaNaNa(E/Z)

CAS RN 192575-19-2

IUPAC Name: tetrasodium 4-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]-2,5-dihydropyridin-2-ylidene}hydrazin-1-yl)benzene-1,3-disulfonate
Synonyms: tetrasodium 4-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]pyridin-2-ylidene}hydrazin-1-yl)benzene-1,3-disulfonate
Molecular Weight (g/mol): 599.3
Molecular Formula: C14H10N3Na4O12PS2
InChi Key: ZNPHTQDEBMCNDG-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].CC1=NC(=NNC2=C(C=C(C=C2)S([O-])(=O)=O)S([O-])(=O)=O)C(COP([O-])([O-])=O)=C(C=O)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products