accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1929-77-7

H3CSONCH3CH3

CAS RN 1929-77-7

IUPAC Name: N,N-dipropyl(propylsulfanyl)formamide
Synonyms: vernolate
Molecular Weight (g/mol): 203.34
Molecular Formula: C10H21NOS
InChi Key: OKUGPJPKMAEJOE-UHFFFAOYSA-N
SMILES: CCCSC(=O)N(CCC)CCC
Concentration 
  • (1)
Element 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products