CAS RN 2095616-82-1
CAS RN 2095616-82-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express CH7057288, MedChemExpress
MedChemExpress CH7057288 is a potent and selective TRK inhibitor.
