accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 2117-28-4

H3CSiCH3H3CSiCH3CH3H3C

CAS RN 2117-28-4

IUPAC Name: trimethyl[(trimethylsilyl)methyl]silane
Synonyms: trimethyl[(trimethylsilyl)methyl]silane
Molecular Weight (g/mol): 160.41
Molecular Formula: C7H20Si2
InChi Key: GYIODRUWWNNGPI-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C[Si](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products