CAS RN 221244-14-0
CAS RN 221244-14-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express 1-NM-PP1, MedChemExpress
MedChemExpress 1-NM-PP1, a cell-permeable PP1 analog, is a potent Src family kinases inhibitor with IC50s of 4.3 nM and 3.2 nM for v-Src-as1 and c-Fyn-as1, respectively.
