accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 222180-19-0

OOHO

CAS RN 222180-19-0

IUPAC Name: 3'-formyl-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 3'-formyl-[1,1'-biphenyl]-3-carboxylic acid
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: QOCUSJPHQSKJJR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CC(C=O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products