CAS RN 230301-11-8
CAS RN 230301-11-8
IUPAC Name:
tert-butyl 1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate
Synonyms:
tert-butyl 1H,4H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate
Molecular Weight (g/mol):
223.28
Molecular Formula:
C11H17N3O2
InChi Key:
BHYPERDTLBCHAE-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1CCC2=C(C1)C=NN2