CAS RN 230615-23-3
CAS RN 230615-23-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Varenicline Hydrochloride, MedChemExpress
MedChemExpress Varenicline Hydrochloride (CP 526555 hydrochloride) is a high affinity, selective α4β2 nicotine acetylcholine receptor (nAChR) partial agonist and full α7 nAChR agonist. Varenicline Hydrochloride is also a potent partial agonist of α6β2 nAChR in striatum of rats with a Ki value of 0.12 nM.
