accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 23228-45-7

FFFClOHO

CAS RN 23228-45-7

IUPAC Name: 2-chloro-4-(trifluoromethyl)benzoic acid
Synonyms: 2-chloro-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: DTIJZEUKFYGSEC-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=C(C=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products