CAS RN 2378173-15-8
CAS RN 2378173-15-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express PF-06928215, MedChemExpress
MedChemExpress PF-06928215 is a cGAS (cyclic GMP-AMP Synthase) inhibitor with an IC50 of 4.9 ÎĽÎĽ. PF-06928215 has a high binding affinity of 0.2 ÎĽM (Kd).
