accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 26878-35-3

HHClClNH2NH2N

CAS RN 26878-35-3

IUPAC Name: dihydrogen pyridine-2,5-diamine dichloride
Synonyms: dihydrogen 2,5-diaminopyridine dichloride
Molecular Weight (g/mol): 182.05
Molecular Formula: C5H9Cl2N3
InChi Key: RWHNUWWUPZKDQP-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)N=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products