CAS RN 2770267-63-3
CAS RN 2770267-63-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ P110

Dynamin-related protein 1 (Drp1) inhibitor; cell-permeable