CAS RN 27832-58-2
CAS RN 27832-58-2
IUPAC Name:
3,3-dimethylpiperidine hydrochloride
Synonyms:
3,3-dimethylpiperidine hydrochloride
Molecular Weight (g/mol):
149.66
Molecular Formula:
C7H16ClN
InChi Key:
BXFKQBZPFQBCCO-UHFFFAOYSA-N
SMILES:
Cl.CC1(C)CCCNC1
1
–
1
of
1
results