accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 28267-29-0

H3COOCH3

CAS RN 28267-29-0

IUPAC Name: ethyl tridecanoate
Synonyms: ethyl tridecanoate
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: QJYYMNOTJXIOBP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC(=O)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products