CAS RN 30123-17-2
CAS RN 30123-17-2
IUPAC Name:
sodium 2-[(6-carboxyhexyl)amino]-6-chloro-10-methyl-9λ⁴-thia-10-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-9,9-bis(olate)
Synonyms:
tianeptine
Molecular Weight (g/mol):
459.94
Molecular Formula:
C21H25ClN2NaO4S
InChi Key:
KBNZQVDKURREAX-UHFFFAOYNA-L
SMILES:
[Na+].CN1C2=CC=CC=C2C(NCCCCCCC(O)=O)C2=CC=C(Cl)C=C2S1([O-])[O-]
1
–
1
of
1
results
1
Thermo Scientific Chemicals Tianeptine, sodium salt
CAS: 30123-17-2 Molecular Formula: C21H25ClN2NaO4S