CAS RN 33252-32-3
CAS RN 33252-32-3
IUPAC Name:
2-amino-4-ethylpyridin-1-ium
Synonyms:
2-amino-4-ethylpyridin-1-ium
Molecular Weight (g/mol):
123.18
Molecular Formula:
C7H11N2
InChi Key:
SJWHILBZPGQBJE-UHFFFAOYSA-O
SMILES:
CCC1=CC=[NH+]C(N)=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Amino-4-ethylpyridine, 97%
CAS: 33252-32-3 Molecular Formula: C7H11N2 Molecular Weight (g/mol): 123.18 MDL Number: MFCD00129025 InChI Key: SJWHILBZPGQBJE-UHFFFAOYSA-O Synonym: 2-amino-4-ethylpyridine,4-ethyl-pyridin-2-ylamine,4-ethyl-2-pyridinamine,2-pyridinamine, 4-ethyl,chembl62077,pubchem7108,4-ethyl-2-pyridylamine,4-ethyl-2-aminopyridine,acmc-20ao52 PubChem CID: 118425 IUPAC Name: 4-ethylpyridin-2-amine SMILES: CCC1=CC=[NH+]C(N)=C1