accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 3344-18-1

MgMgMgHOOOHOHOOHOHOOOHOHOOHO

CAS RN 3344-18-1

IUPAC Name: bis(2-hydroxypropane-1,2,3-tricarboxylic acid) trimagnesium
Synonyms: magnesium citrate (3:2)
Molecular Weight (g/mol): 457.16
Molecular Formula: C12H16Mg3O14
InChi Key: AOHMFUYIHARAGR-UHFFFAOYSA-N
SMILES: [Mg].[Mg].[Mg].OC(=O)CC(O)(CC(O)=O)C(O)=O.OC(=O)CC(O)(CC(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products