CAS RN 351003-36-6
CAS RN 351003-36-6
IUPAC Name:
2-fluoro-5-iodobenzonitrile
Synonyms:
2-fluoro-5-iodobenzonitrile
Molecular Weight (g/mol):
247.01
Molecular Formula:
C7H3FIN
InChi Key:
BIZHQRAAZMDWNK-UHFFFAOYSA-N
SMILES:
FC1=C(C=C(I)C=C1)C#N
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Fluoro-5-iodobenzonitrile, 97%
CAS: 351003-36-6 Molecular Formula: C7H3FIN Molecular Weight (g/mol): 247.01 MDL Number: MFCD03094225 InChI Key: BIZHQRAAZMDWNK-UHFFFAOYSA-N Synonym: 2-fluoro-5-iodobenzonitirile,2-fluoro-5-iodo-benzonitrile,2-fluoro-5-iodobenzonitirle,benzonitrile, 2-fluoro-5-iodo,2-fluoro-5-iodobenzenecarbonitrile,pubchem4779,ksc493m3h,2-fluoro-5-iodobenzonitrile PubChem CID: 2778928 IUPAC Name: 2-fluoro-5-iodobenzonitrile SMILES: FC1=C(C=C(I)C=C1)C#N