accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 3623-15-2

OHC

CAS RN 3623-15-2

IUPAC Name: 1-phenylprop-2-yn-1-one
Synonyms: 1-phenylprop-2-yn-1-one
Molecular Weight (g/mol): 130.15
Molecular Formula: C9H6O
InChi Key: JITPLZPWKYUTDM-UHFFFAOYSA-N
SMILES: O=C(C#C)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products