accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 374564-36-0

BOFFFK

CAS RN 374564-36-0

IUPAC Name: potassium trifluoro(4-formylphenyl)boranuide
Synonyms: potassium trifluoro(4-formylphenyl)boranuide
Molecular Weight (g/mol): 212.02
Molecular Formula: C7H5BF3KO
InChi Key: PEWVRVCGAOIWBB-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(C=O)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products