CAS RN 392-71-2
CAS RN 392-71-2
IUPAC Name:
2,6-dichloro-4-fluorophenol
Synonyms:
2,6-dichloro-4-fluorophenol
Molecular Weight (g/mol):
180.99
Molecular Formula:
C6H3Cl2FO
InChi Key:
BOJVIFKSTRCIRJ-UHFFFAOYSA-N
SMILES:
OC1=C(Cl)C=C(F)C=C1Cl
1
–
2
of
2
results
1
Thermo Scientific Alfa Aesar 2,6-Dichloro-4-fluorophenol, 98%
CAS: 392-71-2 Molecular Formula: C6H3Cl2FO Molecular Weight (g/mol): 180.987 MDL Number: MFCD00010675 InChI Key: BOJVIFKSTRCIRJ-UHFFFAOYSA-N PubChem CID: 94950 IUPAC Name: 2,6-dichloro-4-fluorophenol SMILES: C1=C(C=C(C(=C1Cl)O)Cl)F