accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 40571-86-6

OH(R)(R)H2N

CAS RN 40571-86-6

IUPAC Name: (1R,2R)-N-benzyl-2-hydroxycyclohexan-1-aminium
Synonyms: (1R,2R)-N-benzyl-2-hydroxycyclohexan-1-aminium
Molecular Weight (g/mol): 206.31
Molecular Formula: C13H20NO
InChi Key: NJNCYFUGUYIMEQ-CHWSQXEVSA-O
SMILES: O[C@@H]1CCCC[C@H]1[NH2+]CC1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products