accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 4403-70-7

NH2NH2

CAS RN 4403-70-7

IUPAC Name: 3-(aminomethyl)aniline
Synonyms: 3-(aminomethyl)aniline
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: ZDBWYUOUYNQZBM-UHFFFAOYSA-N
SMILES: NCC1=CC(N)=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products