accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 445-82-9

H3COFOCH3

CAS RN 445-82-9

IUPAC Name: 1-(5-fluoro-2-methoxyphenyl)ethan-1-one
Synonyms: 1-(5-fluoro-2-methoxyphenyl)ethanone
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: CNVGMLMIQIPAFY-UHFFFAOYSA-N
SMILES: COC1=C(C=C(F)C=C1)C(C)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products