accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 459-47-2

H3CF

CAS RN 459-47-2

IUPAC Name: 1-ethyl-4-fluorobenzene
Synonyms: 1-ethyl-4-fluorobenzene
Molecular Weight (g/mol): 124.16
Molecular Formula: C8H9F
InChi Key: BLDNWXVISIXWKZ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(F)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products