CAS RN 459147-39-8
CAS RN 459147-39-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ SW 033291

High affinity 15-PGDH inhibitor; promotes hematopoiesis and hepatocyte proliferation
2
Medchem Express SW033291, MedChemExpress
MedChemExpress SW033291 is a potent and high-affinity inhibitor of 15-PGDH with a Ki of 0.1 nM. SW033291 increases prostaglandin PGE2 levels in bone marrow and other tissues. SW033291 also promotes tissue regeneration.

3
Tocris Bioscience™ SW 033291

High affinity 15-PGDH inhibitor; promotes hematopoiesis and hepatocyte proliferation

4
Tocris Bioscience™ SW 033291

High affinity 15-PGDH inhibitor; promotes hematopoiesis and hepatocyte proliferation
