accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 462-60-2

H2NONHOHO

CAS RN 462-60-2

IUPAC Name: 2-(carbamoylamino)acetic acid
Synonyms: N-(aminocarbonyl)glycine
Molecular Weight (g/mol): 118.09
Molecular Formula: C3H6N2O3
InChi Key: KZVRXPPUJQRGFN-UHFFFAOYSA-N
SMILES: NC(=O)NCC(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products