accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 4621-66-3

H2NSN

CAS RN 4621-66-3

IUPAC Name: pyridine-3-carbothioamide
Synonyms: thionicotinamide
Molecular Weight (g/mol): 138.19
Molecular Formula: C6H6N2S
InChi Key: XQWBMZWDJAZPPX-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CN=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products