CAS RN 466-24-0
CAS RN 466-24-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Benzoylaconine, MedChemExpress
MedChemExpress Benzoylaconine(Isaconitine; Pikraconitin) is an alkaloid in the Chinese traditional medicine Radix Aconiti Lateralis Preparata (Fuzi).
