accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 53600-33-2

H3COOHOH2N

CAS RN 53600-33-2

IUPAC Name: 2-amino-6-methoxybenzoic acid
Synonyms: 2-amino-6-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: DYZDIWNRWSNVPT-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=C1C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products