accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 5396-71-4

CH3OONOO(E)

CAS RN 5396-71-4

IUPAC Name: ethyl (2E)-3-(3-nitrophenyl)prop-2-enoate
Synonyms: ethyl (2E)-3-(3-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol): 221.21
Molecular Formula: C11H11NO4
InChi Key: QZEPRSLOWNHADS-VOTSOKGWSA-N
SMILES: CCOC(=O)\C=C\C1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products