accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 54010-71-8

OOHOHOHOHOPOOHONa

CAS RN 54010-71-8

IUPAC Name: sodium 6-(hydrogen phosphonatooxy)-2,3,4,5-tetrahydroxyhexanal
Synonyms: sodium glucose-6 phosphate
Molecular Weight (g/mol): 282.12
Molecular Formula: C6H12NaO9P
InChi Key: OBHLNVXMRZXIII-UHFFFAOYNA-M
SMILES: [Na+].OC(COP(O)([O-])=O)C(O)C(O)C(O)C=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products