accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 596-51-0

BrCH3NH3COOHO

CAS RN 596-51-0

IUPAC Name: 3-[(2-cyclopentyl-2-hydroxy-2-phenylacetyl)oxy]-1,1-dimethylpyrrolidin-1-ium bromide
Synonyms: glycopyrronium bromide
Molecular Weight (g/mol): 398.34
Molecular Formula: C19H28BrNO3
InChi Key: VPNYRYCIDCJBOM-UHFFFAOYNA-M
SMILES: [Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(C1CCCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products