accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 597-12-6

OH(R)O(R)O(S)(R)OH(R)HOO(S)HOO(R)O(R)HO(S)HO(S)OH(R)OH(R)OH(S)OH(S)OH

CAS RN 597-12-6

IUPAC Name: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2R,3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol melezitose melizitose
Molecular Weight (g/mol): 504.44
Molecular Formula: C18H32O16
InChi Key: QWIZNVHXZXRPDR-WSCXOGSTSA-N
SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products