accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 59768-74-0

CH3CH3CH3OOHNOOH3COHO

CAS RN 59768-74-0

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol): 247.25
Molecular Formula: C10H17NO6
InChi Key: WFPSMPYVXFVVFA-UHFFFAOYNA-N
SMILES: COC(=O)CC(NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products