accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 60207-90-1

CH3ONNNOClCl

CAS RN 60207-90-1

IUPAC Name: 1-{[2-(2,4-dichlorophenyl)-4-propyl-1,3-dioxolan-2-yl]methyl}-1H-1,2,4-triazole
Synonyms: propiconazole radar desmel orbit banner caswell no. 323EE epa pesticide chemical code 122101 Tilt
Molecular Weight (g/mol): 342.22
Molecular Formula: C15H17Cl2N3O2
InChi Key: STJLVHWMYQXCPB-UHFFFAOYNA-N
SMILES: CCCC1COC(CN2C=NC=N2)(O1)C1=CC=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (4)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (3)
Grade 
  • (1)
Industry Type 
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products