CAS RN 613660-87-0
CAS RN 613660-87-0
IUPAC Name:
(4-sulfamoylphenyl)boronic acid
Synonyms:
4-sulfamoylphenylboronic acid
Molecular Weight (g/mol):
201
Molecular Formula:
C6H8BNO4S
InChi Key:
AKSXQPCIAOIJGP-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=CC=C(C=C1)B(O)O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4-Sulfamoylbenzeneboronic acid, 97%, Thermo Scientific™
CAS: 613660-87-0 Molecular Formula: C6H8BNO4S Molecular Weight (g/mol): 201.003 MDL Number: MFCD06659839 InChI Key: AKSXQPCIAOIJGP-UHFFFAOYSA-N Synonym: 4-aminosulfonylphenyl boronic acid,4-aminosulfonyl phenylboronic acid,4-sulfamoylphenyl boronic acid,4-boronobenzenesulfonamide,4-aminosulphonyl benzeneboronic acid,4-aminosulfonyl benzeneboronic acid,4-sulphamoylbenzeneboronic acid,4-sulfamoylbenzeneboronic acid,4-boronobenzenesulphonamide,boronic acid, 4-aminosulfonyl phenyl PubChem CID: 22391977 IUPAC Name: (4-sulfamoylphenyl)boronic acid SMILES: B(C1=CC=C(C=C1)S(=O)(=O)N)(O)O