accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 6240-11-5

OH

CAS RN 6240-11-5

IUPAC Name: 2-(adamantan-1-yl)ethan-1-ol
Synonyms: 2-(adamantan-1-yl)ethanol
Molecular Weight (g/mol): 180.29
Molecular Formula: C12H20O
InChi Key: ZBIDZPHRNBZTLT-UHFFFAOYSA-N
SMILES: OCCC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products