CAS RN 6307-83-1
CAS RN 6307-83-1
IUPAC Name:
3-bromo-5-nitrobenzoate
Synonyms:
3-bromo-5-nitrobenzoate
Molecular Weight (g/mol):
245.01
Molecular Formula:
C7H3BrNO4
InChi Key:
AXRKIZCFYZBBPX-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC(Br)=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 3-Bromo-5-nitrobenzoic acid, 99%, Thermo Scientific™
CAS: 6307-83-1 Molecular Formula: C7H4BrNO4 Molecular Weight (g/mol): 246.016 MDL Number: MFCD00100098 InChI Key: AXRKIZCFYZBBPX-UHFFFAOYSA-N Synonym: 3-bromo-5-nitrobenzoic acid,5-bromo-3-nitrobenzoic acid,benzoic acid, 3-bromo-5-nitro,3-bromo-5-nitro-benzoic acid,3-bromo-5-nitrobenzoicacid,pubchem4734,acmc-209ncw,ksc357s4t,3-bromo-5-nitro benzoic acid PubChem CID: 239336 SMILES: C1=C(C=C(C=C1[N+](=O)[O-])Br)C(=O)O